Stampa…

9-Fluorenone-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 9-Oxofluoren-4-carboxylic acid, 9-Oxo-9H-fluorene-4-carboxylic acid, 9-Oxofluorene-4-carboxylic acid

F0804-1G F0804-5G
TCIAF0804-1GEA 21 EUR
TCIAF0804-1G TCIAF0804-5G
9-Fluorenone-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
9-Fluorenone-4-carboxylic acid
Formula: C₁₄H₈O₃
Peso molecolare: 224,22 g/mol
MDL Number: MFCD00001145
CAS Number: 6223-83-2
EINECS: 228-311-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbb564542384b391e36f1dea8d5eac6f0b044137&vid=fbb564542384b391e36f1dea8d5eac6f0b044137&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 226.0 to 230.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR