Stampa…

9-Fluorenone-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 9-Oxofluoren-1-carboxylic acid

F0590-1G F0590-5G
TCIAF0590-1GEA 31 EUR
TCIAF0590-1G TCIAF0590-5G
9-Fluorenone-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
9-Fluorenone-1-carboxylic acid
Formula: C₁₄H₈O₃
Peso molecolare: 224,22 g/mol
MDL Number: MFCD00011537
CAS Number: 1573-92-8
EINECS: 216-396-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aa9dcb63c9ca99d21e02590c49cc2b5ad4a6d7e&vid=3aa9dcb63c9ca99d21e02590c49cc2b5ad4a6d7e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR