Stampa…

Rosso chinolina ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

Q0007-1G Q0007-25G
TCIAQ0007-1GEA 16 EUR
TCIAQ0007-1G TCIAQ0007-25G
Rosso chinolina ≥95.0% (by titrimetric analysis)
Rosso chinolina
Formula: C₂₁H₂₃IN₂
Peso molecolare: 430,33 g/mol
Punto di fusione: 240 °C
MDL Number: MFCD00011968
CAS Number: 117-92-0
EINECS: 204-221-5
Merck Index: 14,08049
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a759494a820b6172f4c5ca4d511a7f02a77b6608&vid=a759494a820b6172f4c5ca4d511a7f02a77b6608&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Absorbance min. 0.52 (5mg/L methanol, 524 to 528nm)
Transition interval (pH) (colorless to very pale red)1.4 to 3.2 (yellowish red)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR