Nitroterephthalic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Nitroterephthalic acid
N0272-25G
N0272-5G
TCIAN0272-25GEA
210
EUR
TCIAN0272-25G
TCIAN0272-5G
Nitroterephthalic acid ≥98.0% (by GC, titration analysis)
Nitroterephthalic acid
Formula:
C₈H₅NO₆ Peso molecolare: 211,13 g/mol Punto di fusione: 270…272 °C |
MDL Number:
MFCD00007141 CAS Number: 610-29-7 EINECS: 210-217-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...