4-Aminophthalic acid ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A1512-25G
A1512-5G
TCIAA1512-25GEA
144
EUR
TCIAA1512-25G
TCIAA1512-5G
4-Aminophthalic acid ≥95.0% (by titrimetric analysis)
4-Aminophthalic acid
Formula:
C₈H₇NO₄ Peso molecolare: 181,15 g/mol |
MDL Number:
MFCD00013985 CAS Number: 5434-21-9 EINECS: 226-596-4 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 91.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...