Antralina ≥95.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
T1166-10G
T1166-1G
TCIAT1166-10GEA
227
EUR
TCIAT1166-10G
TCIAT1166-1G
Antralina ≥95.0% (by HPLC, titration analysis)
Antralina
Formula:
C₁₄H₁₀O₃ Peso molecolare: 226,23 g/mol Punto di ebollizione: 400 °C (1013 hPa) Punto di fusione: 193,5…195 °C Densità: 1,433 g/cm³ (20 °C) |
MDL Number:
MFCD00053409 CAS Number: 1143-38-0 EINECS: 214-538-0 Merck Index: 12,00723 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...