Methyl-2-chloro-5-nitrobenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Chloro-5-nitrobenzoic acid methyl ester, Methyl 2-chloro-5-nitrobenzenecarboxylate
C1732-25G
C1732-5G
TCIAC1732-25GEA
107
EUR
TCIAC1732-25G
TCIAC1732-5G
Methyl-2-chloro-5-nitrobenzoate ≥98.0% (by GC)
Methyl-2-chloro-5-nitrobenzoate
Formula:
C₈H₆ClNO₄ Peso molecolare: 215,59 g/mol |
MDL Number:
MFCD00184587 CAS Number: 6307-82-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 67.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...