Stampa…

Methyl-2-bromo-5-nitrobenzoate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 2-Bromobenzoic acid methyl ester, Methyl 2-bromo-5-nitrobenzoate

B1916-25G B1916-5G
TCIAB1916-25GEA 40 EUR
TCIAB1916-25G TCIAB1916-5G
Methyl-2-bromo-5-nitrobenzoate ≥98.0% (by GC)
Methyl-2-bromo-5-nitrobenzoate
Formula: C₈H₆BrNO₄
Peso molecolare: 260.04 g/mol
Punto di fusione: 82
MDL Number: MFCD00010867
CAS Number: 6942-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe8884ee87bca812af5d97ce920911b4b1ed1090&vid=fe8884ee87bca812af5d97ce920911b4b1ed1090&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR