Pyridinium dichromate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
PDC
P0931-25G
P0931-500G
P0931-100G
TCIAP0931-25GEA
20
EUR
TCIAP0931-25G
TCIAP0931-500G
TCIAP0931-100G
Pyridinium dichromate ≥98.0% (by titrimetric analysis)
Pyridinium dichromate
Formula:
C₁₀N₁₂Cr₂N₂O₇ Peso molecolare: 532.19 g/mol |
MDL Number:
MFCD00013105 CAS Number: 20039-37-6 EINECS: 243-478-8 UN: 1479 ADR: 5.1 II |
|
Risultati del test di specificazione
Appearance | Orange to Brown powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...