Stampa…
Pyridinium chlorochromate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Pericolo
Synonyms: PCC
P0930-100G P0930-25G P0930-500G
TCIAP0930-100GEA 37 EUR
TCIAP0930-100G TCIAP0930-25G TCIAP0930-500G
Pyridinium chlorochromate ≥98.0% (by titrimetric analysis)
Pyridinium chlorochromate
Formula: C₅H₆ClCrNO₃
Peso molecolare: 215.56 g/mol
Punto di fusione: 205
MDL Number: MFCD00013106
CAS Number: 26299-14-9
EINECS: 247-595-5
UN: 3087
ADR: 5.1 I
Merck Index: 14,07974
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f9c9d8d567003e3f3625455399fad92541865bc&vid=6f9c9d8d567003e3f3625455399fad92541865bc&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Iodometric Titration) min. 98.0 %