2-Chlorophenothiazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Chloro-10H-phenothiazine
C0248-25G
C0248-500G
TCIAC0248-25GEA
30
EUR
TCIAC0248-25G
TCIAC0248-500G
2-Chlorophenothiazine ≥98.0% (by GC)
2-Chlorophenothiazine
Formula:
C₁₂H₈ClNS Peso molecolare: 233,72 g/mol Punto di fusione: 197…202 °C |
MDL Number:
MFCD00005016 CAS Number: 92-39-7 EINECS: 202-152-5 |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 198.0 to 203.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...