2-Aminoisobutyric acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid
A0323-25G
A0323-500G
A0323-100G
TCIAA0323-25GEA
19
EUR
TCIAA0323-25G
TCIAA0323-500G
TCIAA0323-100G
2-Aminoisobutyric acid ≥98.0% (by titrimetric analysis)
2-Aminoisobutyric acid
Formula:
C₄H₉NO₂ Peso molecolare: 103,12 g/mol Punto di ebollizione: 204 °C (1013 hPa) Punto di fusione: 335 °C Densità: 1,278 g/cm³ (20 °C) |
MDL Number:
MFCD00008049 CAS Number: 62-57-7 EINECS: 200-544-0 Merck Index: 14,00445 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...