Stampa…

2-Amino-4-methylbenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Cyano-5-methylaniline

A2448-1G A2448-5G
TCIAA2448-1GEA 76 EUR
TCIAA2448-1G TCIAA2448-5G
2-Amino-4-methylbenzonitrile ≥98.0% (by GC)
2-Amino-4-methylbenzonitrile
Formula: C₈H₈N₂
Peso molecolare: 132,16 g/mol
MDL Number: MFCD00173706
CAS Number: 26830-96-6
EINECS: 248-020-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3267bc248529dc22359076a3743a74872732318e&vid=3267bc248529dc22359076a3743a74872732318e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR