Stampa…

2-Hydroxyisobutyric acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

M0360-25G M0360-500G M0360-100G
TCIAM0360-25GEA 20 EUR
TCIAM0360-25G TCIAM0360-500G TCIAM0360-100G
2-Hydroxyisobutyric acid ≥98.0% (by titrimetric analysis)
2-Hydroxyisobutyric acid
Formula: C₄H₈O₃
Punto di ebollizione: 212°C
Punto di fusione: 77 °C
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f110a4cd792bf2c170873ee2f57959a51eae4744&vid=f110a4cd792bf2c170873ee2f57959a51eae4744&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 82.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR