Stampa…

2-Nitro-p-toluonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Methyl-2-nitrobenzonitrile

N0494-1G N0494-5G
TCIAN0494-1GEA 84 EUR
TCIAN0494-1G TCIAN0494-5G
2-Nitro-p-toluonitrile ≥98.0% (by GC)
2-Nitro-p-toluonitrile
Formula: C₈H₆N₂O₂
Peso molecolare: 162,15 g/mol
MDL Number: MFCD00056107
CAS Number: 26830-95-5
EINECS: 248-019-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=457c33b35706b60abda7352d2bb27dd88c1f310a&vid=457c33b35706b60abda7352d2bb27dd88c1f310a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR