Stampa…

Bis(2-nitrophenyl)disulphide ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: Bis(2-nitrophenyl) disulphide

B0503-25G B0503-500G
TCIAB0503-25GEA 30 EUR
TCIAB0503-25G TCIAB0503-500G
Bis(2-nitrophenyl)disulphide ≥98.0% (by GC, total nitrogen)
Bis(2-nitrophenyl)disulphide
Formula: C₁₂H₈N₂O₄S₂
Peso molecolare: 308,34 g/mol
Punto di fusione: 198 °C
Densità: 1,625 g/cm³ (-123.15 °C)
MDL Number: MFCD00007130
CAS Number: 1155-00-6
EINECS: 214-581-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=628c63a608cc9e7eeb89301eb7c064ff76b5911c&vid=628c63a608cc9e7eeb89301eb7c064ff76b5911c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Acetic acid very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR