Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Sodium 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic acid sodium salt
C0991-25G
C0991-500G
C0991-100G
TCIAC0991-25GEA
23
EUR
TCIAC0991-25G
TCIAC0991-500G
TCIAC0991-100G
Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)
Sodium chlorodifluoroacetate
Formula:
C₂ClF₂NaO₂ Peso molecolare: 152,46 g/mol Punto di fusione: 193…195 °C |
MDL Number:
MFCD00064771 CAS Number: 1895-39-2 EINECS: 217-586-0 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...