Stampa…

6-Methylnicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

M1894-25G M1894-5G
TCIAM1894-25GEA 282 EUR
TCIAM1894-25G TCIAM1894-5G
6-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
6-Methylnicotinic acid
Formula: C₇H₇NO₂
Peso molecolare: 137,14 g/mol
Punto di fusione: 210°C
MDL Number: MFCD00006341
CAS Number: 3222-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8e9c47638b73ebcf0e20f54eb9cc7c749d4ebb3&vid=a8e9c47638b73ebcf0e20f54eb9cc7c749d4ebb3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR