Stampa…

5-Chloronicotinic acid ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C2399-1G C2399-5G
TCIAC2399-1GEA 44 EUR
TCIAC2399-1G TCIAC2399-5G
5-Chloronicotinic acid ≥95.0% (by GC, titration analysis)
5-Chloronicotinic acid
Formula: C₆H₄ClNO₂
MDL Number: MFCD00128901
CAS Number: 22620-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d05fc3f76d790b85ecb339ec41d31cf0ba031dff&vid=d05fc3f76d790b85ecb339ec41d31cf0ba031dff&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR