Cytosine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol
C0528-25G
C0528-5G
C0528-100G
TCIAC0528-25GEA
36
EUR
TCIAC0528-25G
TCIAC0528-5G
TCIAC0528-100G
Cytosine ≥98.0% (by HPLC, titration analysis)
Cytosine
Formula:
C₄H₅N₃O Peso molecolare: 111,1 g/mol Punto di fusione: 313…316 °C Densità: 0,99771 g/cm³ (25 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006034 CAS Number: 71-30-7 EINECS: 200-749-5 Merck Index: 14,02795 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in dilute HCl | almost transparency |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...