Stampa…

1-Ethyl-3-methylimidazolium dicyanamide ≥97.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: EMIM DCA

E0680-1G E0680-5G
TCIAE0680-1GEA 25 EUR
TCIAE0680-1G TCIAE0680-5G
1-Ethyl-3-methylimidazolium dicyanamide ≥97.0% (by HPLC, total nitrogen)
1-Ethyl-3-methylimidazolium dicyanamide
Formula: C₈H₁₁N₅
Peso molecolare: 177,21 g/mol
Flash Pt: 221°C
MDL Number: MFCD08276373
CAS Number: 370865-89-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47814f6a15c0e6a2b80e4f8b904f7942c4d8c849&vid=6c0d25cf7fe002c8733e451a73aba00931201825&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 97.0 % (total of Ion-Pair)
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR