Stampa…

2-Amino-m-cresol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

A1851-25G A1851-5G
TCIAA1851-25GEA 335 EUR
TCIAA1851-25G TCIAA1851-5G
2-Amino-m-cresol ≥98.0% (by GC, titration analysis)
2-Amino-m-cresol
Formula: C₇H₉NO
Peso molecolare: 123,15 g/mol
MDL Number: MFCD00075082
CAS Number: 2835-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bdf4cbefdb595ae2907e7c8b481c4a9c40f8128&vid=0bdf4cbefdb595ae2907e7c8b481c4a9c40f8128&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR