Stampa…

N,N-Diethylethylenediamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(Diethylamino)ethylamine, 2-Aminoethyldiethylamine

D0505-25ML D0505-500ML
TCIAD0505-25MLEA 18 EUR
TCIAD0505-25ML TCIAD0505-500ML
N,N-Diethylethylenediamine ≥98.0% (by GC, titration analysis)
N,N-Diethylethylenediamine
Formula: C₆H₁₆N₂
Peso molecolare: 116,21 g/mol
Punto di ebollizione: 146°C
Punto di fusione: –70 °C
Densità: 0,82 g/cm³ (20 °C)
Flash Pt: 40°C
MDL Number: MFCD00008176
CAS Number: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e428e7cdd3c8d6a47a712233910569b782c259&vid=a0e428e7cdd3c8d6a47a712233910569b782c259&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR