4-Nitro-2,6-xylenol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₉NO₃ |
MDL Number:
MFCD00007339 CAS Number: 2423-71-4 EINECS: 219-353-9 |
|
Risultati del test di specificazione
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...