Trimethylsulphonium bromide ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₃H₉BrS Punto di fusione: 203 °C Densità: 1,67 g/cm³ (20 °C) |
MDL Number:
MFCD00051367 CAS Number: 3084-53-5 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...