trans-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
C0474-25G
C0474-500G
TCIAC0474-25GEA
91
EUR
TCIAC0474-25G
TCIAC0474-500G
trans-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
trans-1,2-Cyclohexanedicarboxylic acid
Formula:
C₈H₁₂O₄ Punto di ebollizione: 339 °C (1013 hPa) Punto di fusione: 220…221 °C |
MDL Number:
MFCD00001464 CAS Number: 2305-32-0 EINECS: 218-975-8 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 225.0 to 232.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...