Benzyl propionate ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Propionic acid benzyl ester, Benzyl propanoate
P0501-25ML
P0501-500ML
TCIAP0501-25MLEA
17
EUR
TCIAP0501-25ML
TCIAP0501-500ML
Benzyl propionate ≥99.0% (by GC)
Benzyl propionate
Formula:
C₁₀H₁₂O₂ Peso molecolare: 164,2 g/mol Punto di ebollizione: 222°C Densità: 1,029 to 1.033 g/cm³ (20 °C) Flash Pt: 108°C |
MDL Number:
MFCD00027003 CAS Number: 122-63-4 EINECS: 204-559-3 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...