N,N,N-Trimethylanilinium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Phenyltrimethylammonium bromide, Trimethylphenylammonium bromide
Formula:
C₉H₁₄BrN Peso molecolare: 216.12 g/mol Punto di fusione: 214 |
MDL Number:
MFCD00011788 CAS Number: 16056-11-4 EINECS: 240-202-8 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...