Phenoxyacetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2-Phenoxyacetic acid
P0107-25G
P0107-500G
TCIAP0107-25GEA
16
EUR
TCIAP0107-25G
TCIAP0107-500G
Phenoxyacetic acid ≥98.0% (by GC, titration analysis)
Phenoxyacetic acid
Formula:
C₈H₈O₃ Peso molecolare: 152.15 g/mol Punto di ebollizione: 285°C |
MDL Number:
MFCD00004296 CAS Number: 122-59-8 EINECS: 204-556-7 Merck Index: 14,07254 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 99.0 to 102.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...