Stampa…

Octyltrichlorosilane ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

O0168-25G O0168-250G
TCIAO0168-25GEA 136 EUR
TCIAO0168-25G TCIAO0168-250G
Octyltrichlorosilane ≥98.0% (by GC, titration analysis)
Octyltrichlorosilane
Formula: C₈H₁₇Cl₃Si
Punto di ebollizione: 229°C
Densità: 1,07 g/cm³ (20 °C)
Flash Pt: 96°C
MDL Number: MFCD00000488
CAS Number: 5283-66-9
EINECS: 226-112-1
UN: 1801
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e1d99b739f825884a20fbd096d49f451a77f1d&vid=63e1d99b739f825884a20fbd096d49f451a77f1d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR