Methoxytrimethylsilane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Trimethyl(methoxy)silane
M0688-25ML
M0688-250ML
TCIAM0688-25MLEA
24
EUR
TCIAM0688-25ML
TCIAM0688-250ML
Methoxytrimethylsilane ≥98.0% (by GC)
Methoxytrimethylsilane
Formula:
C₄H₁₂OSi Peso molecolare: 104,22 g/mol Punto di ebollizione: 57°C Densità: 0,755 g/cm³ (20 °C) Flash Pt: –30°C |
MDL Number:
MFCD00008344 CAS Number: 1825-61-2 EINECS: 217-369-0 UN: 1993 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...