Stampa…

N,O-Bis(trimetilsilil)acetammide ≥80.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

B0511-10ML B0511-100ML
TCIAB0511-10MLEA 17 EUR
TCIAB0511-10ML TCIAB0511-100ML
N,O-Bis(trimetilsilil)acetammide ≥80.0% (by GC)
N,O-Bis(trimetilsilil)acetammide
Formula: C₈H₂₁NOSi₂
Peso molecolare: 203,43 g/mol
Punto di ebollizione: 73°C
Punto di fusione: ∼24 °C
Densità: 0,82 g/cm³ (20 °C)
Flash Pt: 42°C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=398d83b7a7918ced1e71e5c5e72278108f80ee5a&vid=398d83b7a7918ced1e71e5c5e72278108f80ee5a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 80.0 %
Effect test of TMS derivatization Effective as derivatization agent DL-Threonine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR