Dimethyl sulphone ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl sulphone, Dimethylsulphone
M0509-25G
M0509-500G
TCIAM0509-25GEA
16
EUR
TCIAM0509-25G
TCIAM0509-500G
Dimethyl sulphone ≥99.0% (by GC)
Dimethyl sulphone
Formula:
C₂H₆O₂S Peso molecolare: 94,13 g/mol Punto di ebollizione: 238°C Punto di fusione: 107…110 °C Densità: 1,17 g/cm³ (110 °C) |
MDL Number:
MFCD00007566 CAS Number: 67-71-0 EINECS: 200-665-9 Merck Index: 14,03258 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 108.0 to 112.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...