N-Methylmaleimide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-Methyl-1H-pyrrole-2,5-dione
M0807-25G
M0807-500G
M0807-100G
TCIAM0807-25GEA
103
EUR
TCIAM0807-25G
TCIAM0807-500G
TCIAM0807-100G
N-Methylmaleimide ≥98.0% (by GC)
N-Methylmaleimide
Formula:
C₅H₅NO₂ Peso molecolare: 111,1 g/mol Punto di ebollizione: 194 °C (1013 hPa) Punto di fusione: 70…75 °C |
MDL Number:
MFCD00005508 CAS Number: 930-88-1 EINECS: 213-226-1 UN: 1759 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 98.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...