Stampa…

Methyl bromoacetate ≥98.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Bromoacetic methyl ester

B0533-25G B0533-500G B0533-100G
TCIAB0533-25GEA 16 EUR
TCIAB0533-25G TCIAB0533-500G TCIAB0533-100G
Methyl bromoacetate ≥98.0% (by GC)
Methyl bromoacetate
Formula: BrCH₂COOCH₃
Peso molecolare: 152.98 g/mol
Punto di ebollizione: 144°C
Punto di fusione: -50
Flash Pt: 63°C
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&vid=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR