Stampa…

8-Bromocaprylic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: 8-Bromooctanoic acid, ω-Bromocaprylic acid, 8-Bromooctoic acid, ω-Bromooctoic acid, ω-Bromooctanoic acid

B1675-25G B1675-5G
TCIAB1675-25GEA 103 EUR
TCIAB1675-25G TCIAB1675-5G
8-Bromocaprylic acid ≥97.0% (by titrimetric analysis)
8-Bromocaprylic acid
Formula: BrH₂C(CH₂)₆COOH
Peso molecolare: 223.11 g/mol
Punto di ebollizione: 150°C
Punto di fusione: 38.5
Densità: 1.324
MDL Number: MFCD00004430
CAS Number: 17696-11-6
UN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae052de0ddbee49b9b259e773965d57fc17458bd&vid=ae052de0ddbee49b9b259e773965d57fc17458bd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 35.0 to 39.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR