Stampa…

6-Nitro-1H-indazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Nitroindazole, 6-Nitro-1H-indazole, 6-NINA

N0399-25G N0399-250G
TCIAN0399-25GEA 21 EUR
TCIAN0399-25G TCIAN0399-250G
6-Nitro-1H-indazole ≥98.0% (by GC)
6-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
Peso molecolare: 163,14 g/mol
Punto di fusione: 181 °C
MDL Number: MFCD00005695
CAS Number: 7597-18-4
EINECS: 231-500-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac478aeac5eb4df4b6fea979e4165966487caad9&vid=ac478aeac5eb4df4b6fea979e4165966487caad9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Melting point 180.0 to 184.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR