Stampa…

Indole-6-carbaldehyd ≥98.0% (by GC)

Fornitore: TCI
Attenzione

I0743-1G I0743-5G
TCIAI0743-1GEA 81 EUR
TCIAI0743-1G TCIAI0743-5G
Indole-6-carbaldehyd ≥98.0% (by GC)
Indole-6-carbaldehyd
Formula: C₉H₇NO
MDL Number: MFCD02179596
CAS Number: 1196-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef03959955b74768c633b35143e88aa5e42fbbdc&vid=ef03959955b74768c633b35143e88aa5e42fbbdc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR