5-Phenyl-1H-tetrazole ≥99.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₆N₄ Punto di ebollizione: 331 °C (1013 hPa) Punto di fusione: 222 °C |
MDL Number:
MFCD00022388 CAS Number: 18039-42-4 EINECS: 241-950-8 UN: 1325 ADR: 4.1,III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...