5-Fluoroindole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉H₆FNO₂ Punto di fusione: 249°C |
MDL Number:
MFCD00005612 CAS Number: 399-76-8 EINECS: 206-919-5 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...