5-Chlorouracil ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
5-Chloropyrimidine-2,4(1H,3H)-dione, Chlorouracil
C0969-25G
C0969-5G
TCIAC0969-25GEA
83
EUR
TCIAC0969-25G
TCIAC0969-5G
5-Chlorouracil ≥98.0% (by titrimetric analysis)
5-Chlorouracil
Formula:
C₄H₃ClN₂O₂ Peso molecolare: 146,53 g/mol Punto di fusione: > 300 °C |
MDL Number:
MFCD00006019 CAS Number: 1820-81-1 EINECS: 217-339-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in NH3aq. | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...