Stampa…

5-Amino-1H-indazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine

A0797-25G A0797-5G
TCIAA0797-25GEA 223 EUR
TCIAA0797-25G TCIAA0797-5G
5-Amino-1H-indazole ≥98.0% (by HPLC, titration analysis)
5-Amino-1H-indazole
Formula: C₇H₇N₃
Peso molecolare: 133,15 g/mol
Punto di ebollizione: 345 °C (1013 hPa)
Punto di fusione: 183 °C
MDL Number: MFCD00037975
CAS Number: 19335-11-6
EINECS: 242-971-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a35335b28ae078ca851ab1c6f8106a2751d2123&vid=8a35335b28ae078ca851ab1c6f8106a2751d2123&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 176.0 to 181.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR