Levulinic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid
L0042-25G
L0042-500G
TCIAL0042-25GEA
17
EUR
TCIAL0042-25G
TCIAL0042-500G
Levulinic acid ≥97.0% (by GC, titration analysis)
Levulinic acid
Formula:
C₅H₈O₃ Peso molecolare: 116,12 g/mol Punto di ebollizione: 245 °C (1013 hPa) Punto di fusione: 26…29 °C Densità: 1,14 g/cm³ (20 °C) Flash Pt: 138 °C |
MDL Number:
MFCD00002796 CAS Number: 123-76-2 EINECS: 204-649-2 UN: 3261 ADR: 8,III Merck Index: 14,05472 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...