Stampa…

4-Nitrophthalimide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Nitroisoindoline-1,3-dione

N0247-25G N0247-500G
TCIAN0247-25GEA 19 EUR
TCIAN0247-25G TCIAN0247-500G
4-Nitrophthalimide ≥98.0% (by HPLC, titration analysis)
4-Nitrophthalimide
Formula: C₈H₄N₂O₄
Peso molecolare: 192,13 g/mol
Punto di fusione: 215 °C
MDL Number: MFCD00005884
CAS Number: 89-40-7
EINECS: 201-905-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49622268bc21b10fb7ab9f28816d3790914488ad&vid=49622268bc21b10fb7ab9f28816d3790914488ad&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR