4-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
N0244-25G
N0244-500G
TCIAN0244-25GEA
27
EUR
TCIAN0244-25G
TCIAN0244-500G
4-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
4-Nitrophthalic acid
Formula:
C₈H₅NO₆ Peso molecolare: 211,13 g/mol Punto di fusione: 162…165 °C |
MDL Number:
MFCD00007252 CAS Number: 610-27-5 EINECS: 210-215-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...