Stampa…

Ethyl-p-toluate ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 4-methylbenzoate, Ethyl-4-methylbenzoate, 4-Methylbenzoic acid ethyl ester, 4-Toluic acid ethyl ester, p-Toluic acid ethyl ester

T0295-25G T0295-500G
TCIAT0295-25GEA 21 EUR
TCIAT0295-25G TCIAT0295-500G
Ethyl-p-toluate ≥99.0% (by GC)
Ethyl-p-toluate
Formula: C₁₀H₁₂O₂
Peso molecolare: 164,2 g/mol
Punto di ebollizione: 232°C
Densità: 1,03 g/cm³ (20 °C)
Flash Pt: 118°C
MDL Number: MFCD00009117
CAS Number: 94-08-6
EINECS: 202-301-4
Merck Index: 14,03840
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9a1c19acfc3be7ea5b1e076289e586fb37ce0bc&vid=e9a1c19acfc3be7ea5b1e076289e586fb37ce0bc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR