4-Iodopyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
I0673-25G
I0673-5G
TCIAI0673-25GEA
254
EUR
TCIAI0673-25G
TCIAI0673-5G
4-Iodopyridine ≥98.0% (by GC, titration analysis)
4-Iodopyridine
Formula:
C₅H₄IN Peso molecolare: 205 g/mol Punto di fusione: 108°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD02093937 CAS Number: 15854-87-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...