Indole-4-carbaldehyd ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1H-Indole-4-carbaldehyde, Indole-4-carboxaldehyde
I0712-1G
I0712-5G
TCIAI0712-1GEA
71
EUR
TCIAI0712-1G
TCIAI0712-5G
Indole-4-carbaldehyd ≥98.0% (by GC)
Indole-4-carbaldehyd
Formula:
C₉H₇NO Peso molecolare: 145,16 g/mol |
MDL Number:
MFCD01632221 CAS Number: 1074-86-8 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 140.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...