Stampa…

2-Thiouracil ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

T0224-25G T0224-250G
TCIAT0224-25GEA 24 EUR
TCIAT0224-25G TCIAT0224-250G
2-Thiouracil ≥98.0% (by HPLC, titration analysis)
2-Thiouracil
Formula: C₄H₄N₂OS
Peso molecolare: 128,15 g/mol
Punto di fusione: 331°C
MDL Number: MFCD00006039
CAS Number: 141-90-2
EINECS: 205-508-8
UN: 3077
ADR: 9,III
Merck Index: 14,09366
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b589923ef4ed909c7be2e1b6de392c09bb79fdeb&vid=b589923ef4ed909c7be2e1b6de392c09bb79fdeb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Sodium hydroxide solution almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR