Stampa…

4-Bromopyridinium chloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

B0653-25G B0653-5G
TCIAB0653-25GEA 140 EUR
TCIAB0653-25G TCIAB0653-5G
4-Bromopyridinium chloride ≥98.0% (by titrimetric analysis)
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
Peso molecolare: 194,46 g/mol
Punto di fusione: 235…238 °C
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a6102314bfa63afac590d5be7fa8a271cd8968&vid=41a6102314bfa63afac590d5be7fa8a271cd8968&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR